Gas calibration standards
Catalog # REST34457
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:Argon, 99,9% product grade, 14 l
- Seq:05
Specifications
About this item
These transportable pure gases and mixtures are meant for purging or calibrating to multi component mixes, which are ideal for peak identification work.
The 14 l container has a CGA 160 connection for more precise integration with analytical systems and 48 l cylinder has a CGA 165 connection and can deliver large volumes of sample.