Transfection reagents, DharmaFECT® Duo
Supplier: Horizon Discovery
Certificates
About this item
A lipid-based reagent specifically formulated for co-transfection of plasmid and siRNA.
- Effective delivery of two distinct molecules with a single formulation
- High transfection efficiency of both siRNA and plasmid without compromising cell viability
- Potent target knockdown and plasmid expression
- Free of microbial contamination and ribonucleases
Specifications
- DNA and siRNA:Yes