Klemmer med fire tænger, allround
Catalog # RSGA470.000
Supplier: RSG
Specifications
- Spændvidde:90 mm
- Pakn:1
Specifications
About this item
Universal clamp made from electrolytically polished stainless steel.
- 4-finger jaws
- Jaws fitted with cork