Order Entry
Denmark
ContactUsLinkComponent
Nut for terminal fitting
Nut for terminal fitting
Catalog # REST24896
Supplier:  Restek
undefined
Nut for terminal fitting
Catalog # REST24896
Supplier:  Restek
Specifications
  • Description:
    Nut for terminal fitting
  • Pk:
    2
Specifications
About this item
These nuts are ideal for use with Thermo Scientific TRACE GCs.