Rods, borosilicate glass
Leverandør: HILGENBERG
Certifikater
Om denne vare
Glass rods are made of borosilicate glass 3.3.
- Very good chemical resistance
- Very good temperature resistance
- Length: 100 mm
With ends cut or fixing of samples and filling of mark-tubes.
Frequently Bought Together