0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vials
Inserts for Versa Vials™
Inserts for Versa Vials™
Inserts for Versa Vials™
Catalog # REST22707
Supplier:
Restek
Expand image
Inserts for Versa Vials™
Catalog # REST22707
Supplier:
Restek
Specifications
Cap included:
No
Capacity:
0,25 ml
Description:
Inserts with glass flange
Material:
Glass
Pk:
100
Specifications
About this item
Product Details
Product Details