Ferrules, Alumaseal®
Supplier: Restek
Certificates
About this item
These ferrules are made of aluminium eliminate out-gassing and make leak-tight seals for less detector noise. Its unique two-piece design permanently locks on fused silica tubing without causing breakage.
- No retightening required after temperature cycles
- Will not crack or fragment
- Will not stick in fittings
- Use with any 1,6 mm compression-type fitting
- Stable up to 550 °C
Frequently Bought Together