PTFE ferrules
Supplier: Restek
Certificates
About this item
- Upper temperature limit 250 °C
- 100% PTFE; completely inert
- One-piece design requires no back ferrule
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Dragendorff's reagent, spray reagent for TLC, Supelco®
Supplier: Merck
CERT TOC LL <10PPB FOR 40ML GLASS VIALS 1 * 80 items
Supplier: PMT
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
Microvolume syringes 10 µl
Supplier: Trajan Scientific and Medical
Consumables for Agilent Technologies autosamplers
Supplier: Trajan Scientific and Medical