Order Entry
Denmark
ContactUsLinkComponent
 
Drum filling station
  CEMG8674
 :  CEMO
undefined
Drum filling station
  CEMG8674
 :  CEMO

 

  • Weight:
    46 kg
  • Description:
    PE filling station 450 l
  • W×D×H:
    1350×1380×640 mm
  • Pk:
    1

 

 

This filling station is suitable as a storage and filling station for two 200 L barrels and with additional drum pallet for four 200 L drums.

  • Made from high-quality polyethylene
  • High chemical resistance
  • Can be driven under
  • Collection volume: 450 L