Order Entry
Denmark
ContactUsLinkComponent
Epidermal growth factor (EGF) from E.coli, MP Biomedicals
Epidermal growth factor (EGF) from E.coli, MP Biomedicals
Katalog # ICNA0215457180
Supplier:  MP Biomedicals
CAS Number:  
Epidermal growth factor (EGF) from E.coli, MP Biomedicals
Katalog # ICNA0215457180
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Description:
    EGF, recombinant

Specifications

About this item

Epidermal Growth Factor (EGF) is a polypeptide growth factor which stimulates the proliferation of a wide range of epidermal and epithelial cells.

  • Potent and cost effective mitogen
  • Soluble in water and most aqueous buffers

Produced in a patented E.coli expression system free of animal-derived components.