Tubing for rotarus® peristaltic pumps, Tygon® A-60-G
These tubes are used for industrial applications for its bending fatigue strength, good abrasion resistance and low gas permeability when compared to rubber tubes.
- Temperature resistance from –60 to 135 °C
- Hardness (Shore durometer A): 61
Frequently Bought Together