Trichoderma viride cellulase Y-C, MP Biomedicals
ICNA08320961
:
- Pk:10 g
- CAS No.:9012-54-8
- Species:Trichoderma viride
- Storage conditions:2...8 °C
- Form:Light brown powder
- Enzyme name:Cellulase Y-C
- Enzyme activity:16000 u/g
- pH:4,0 - 5,0
Cellulase Y-C removes cell walls from plant tissues in shorter incubation period without loss of biological activity of the materials.
- Readily soluble in water
- Ideal for protoplast preparation
It is an effective enzyme for use with pectolyase Y-23 for plant cell wall removal. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It has also been used in a study to investigate the liquefaction of hydrothermally pretreated wheat straw at high-solids content.