Order Entry
Denmark
ContactUsLinkComponent
Utility clamps, three pronged
Utility clamps, three pronged
Catalog # RLAA1029
Supplier:  R L ENTERPRISES
undefined
Utility clamps, three pronged
Catalog # RLAA1029
Supplier:  R L ENTERPRISES

Specifications

  • Spanning width:
    90 mm
  • Description:
    Utility clamp, 3-pronged
  • Pk:
    1

Specifications

About this item

This 3-pronged clamp made of diecast aluminium alloy features plastic coated jaws.

  • With screwed shaft and nickel-plated bosshead
  • Plastic coated jaws are chemical resistant
  • Can be screwed up and down shaft 35 mm and locked in any position with a wing nut

The bosshead accepts rods up to 13 mm, which are secured with a stainless steel thumb screw.