Order Entry
Denmark
Orders LinkContactUsLinkComponent
PLOT columns, MXT® metal
PLOT columns, MXT® metal
Supplier:   Restek
PLOT columns, MXT® metal
Supplier:  Restek
Certificates
PLOT columns, MXT® metal
PLOT columns, MXT® metal
12 Options Available Below
Product Details & Documents
PLOT columns are made from Siltek® treated stainless steel and have the same characteristics and performance as fused silica PLOT columns but useful for GCs and other field applications.
  • Made into small coil diameters and perfect for tight spaces
  • Rugged material withstands rough handling and shock
  • Available in 3,5" coil diameter or 7" diameter with 11-pin cage
Product Family Options
MXT®-Msieve 5A
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 3,5" coil
    Capillary
    MXT®-Msieve 5A
    20,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • REST79717
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Msieve 5A
    20,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • 3,5" coil
    Capillary
    MXT®-Msieve 5A
    50,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 19.030,00 kr
    19.030,00 kr
    1 * 1 Items
       19.030,00 kr
  • REST79723
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Msieve 5A
    50,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 13.310,00 kr
    13.310,00 kr
    1 * 1 Items
       13.310,00 kr
MXT®-Alumina BOND/Na2SO4
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • REST79714
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Alumina BOND/Na2SO4
    10,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.700,00 kr
    8.700,00 kr
    1 * 1 Items
       8.700,00 kr
  • 3,5" coil
    Capillary
    MXT®-Alumina BOND/Na2SO4
    10,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.440,00 kr
    12.440,00 kr
    1 * 1 Items
       12.440,00 kr
MXT®-Q-BOND
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • REST79718
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Q-BOND
    8,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.640,00 kr
    6.640,00 kr
    1 * 1 Items
       6.640,00 kr
  • 3,5" coil
    Capillary
    MXT®-Q-BOND
    8,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.500,00 kr
    9.500,00 kr
    1 * 1 Items
       9.500,00 kr
  • REST79716
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Q-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.280,00 kr
    9.280,00 kr
    1 * 1 Items
       9.280,00 kr
  • 3,5" coil
    Capillary
    MXT®-Q-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 13.260,00 kr
    13.260,00 kr
    1 * 1 Items
       13.260,00 kr
MXT®-S-BOND
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 3,5" coil
    Capillary
    MXT®-S-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • REST79712
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-S-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items