Order Entry
Denmark
ContactUsLinkComponent
Restriction enzymes, BamHI, HC, Fermentas
Restriction enzymes, BamHI, HC, Fermentas
Catalog # ER0053
CAS Number:  
undefined
Restriction enzymes, BamHI, HC, Fermentas
Catalog # ER0053
CAS Number:  

Specifications

  • Concentration:
    50 U/µl
  • Pk:
    1 KIT
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    5' overhang
  • Description:
    BamHI, HC
  • Enzyme name:
    BamHI
  • Units:
    2000 units

Specifications

About this item

BamHI restriction enzyme recognises G^GATCC sites and cuts best at 37 °C in its own unique buffer.

  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers