Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sealed Cuvette
Sealed Cuvette
Rectangular sealed liquid cell, 0,1 mm
Katalog # PERCL1270954
Leverandør:   PerkinElmer
Sealed Cuvette
Katalog # PERCL1270954
Leverandør:  PerkinElmer
Sealed Cuvette
Rectangular sealed liquid cell, 0,1 mm
Price
8.230,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Rectangular cuvette filled by potassium bromide (KBr) for a wider spectral range than sodium chloride (NaCl) but more hygroscopic.
Specifikationer
  • Path length:
    0,1 mm
  • Description:
    Rectangular sealed liquid cell