Order Entry
Denmark
ContactUsLinkComponent
Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals
Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals
Catalog # ICNA0215352610
Supplier:  MP Biomedicals
CAS Number:  
Arthrobacter luteus Yeast lytic enzyme, MP Biomedicals
Catalog # ICNA0215352610
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    10 KU
  • Species:
    Arthrobacter luteus
  • Seq:
    01
  • Storage conditions:
    Store at −20 °C
  • Form:
    Tan powder
  • Enzyme name:
    Yeast lytic enzyme
  • Enzyme activity:
    20,000 units/g
  • Enzyme synonyms:
    Lyticase
  • pH:
    7,5

Specifications

About this item

Yeast lytic enzyme is used to digest the cell wall of active yeast cells. It is prepared from a submerged culture of Arthrobacter luteus which effectively lyses cell walls of viable yeast cell.

Lyophilized enzyme partially purified by affinity chromatography
Hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan

The primary enzyme in this preparation responsible for lysis of viable yeast cells is β-1,3-glucan laminaripentaohydrolase. It hydrolyzes glucose polymers at the β-1,3-glucan linkages releasing laminaripentaose as the principal product.

Unit definition: One unit will produce a delta A800 of 0,001 per minute at pH 7,5 and 25 °C, using a suspension of brewers yeast as a substrate in a 3 ml reaction mixture.