Adapter ferrules, Valco®
Supplier: Restek
Certificates
About this item
Fused silica adapter ferrules are made from valcon polyimide, a unique graphite reinforced composite that is specially made to maximise mechanical stability at temperatures up to 350 °C.
Frequently Bought Together