Paper Bags with Woof for Sterilisation
KING626382/00
- Beskrivelse:Paper bag with woof
- B×D×H:380×510×125 mm
- Pakn:250
These paper bags are used for economic packages where the contents of the pack should not be identifiable.
: EN868-1 and EN868-4