Inserts for crimp top and screw thread vials
: Restek
These inserts are used with 2 ml, 11 mm crimp top and 2 ml, 9 mm short cap, screw thread vials.
Note: Big Mouth insert with glass flange is not to be used with 9 mm screw thread vials.
Frequently Bought Together