Guard/retention gap columns, hydroguard® water-resistant (fused silica)
Supplier: Restek
Certificates
About this item
Extend analytical column lifetime by preventing degradation from harsh 'steam-cleaning' water injections.
- Tested with a comprehensive test mix, to ensure high inertness
Maximum temperature: 325 °C
Frequently Bought Together