AlphaTec® 37-510/37-646 General Purpose Gloves
: Ansell
Unlined nitrile gloves especially designed for use in the food industry.
- High performance nitrile compound for enhanced flexibility and dexterity, even in cold environments
- Ansell UltraGrip™ technology: thanks to the raised diamond grip pattern, these gloves offer unmatched grip in wet and fatty conditions
- Latex free formulation: no risk of Type I allergies
- Unflocked: to prevent the risk of food contamination
- Offer superior mechanical properties over natural rubber gloves
Ideal for use in food preparation, processing and handling, meat and poultry processing, fish processing and cleaning.
: EN 388:2016 (2001X), EN ISO 374-1 Type B (JKL), EN ISO 374-5, EN 421; Cat. III.
Food contact approved.
Food contact approved.
- Catalog No:
- ANSE37-646/12
- ANSE37-510/9
- ANSE37-646/9
- Model:
- 37-646
- 37-510
- 37-646
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Raised diamond
- Raised diamond
- Raised diamond
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (2001X), EN ISO 374-1 Type B (JKL), EN ISO 374-5, EN 421
- EN 388:2016 (2001X), EN ISO 374-1 Type B (JKL), EN ISO 374-5, EN 421
- EN 388:2016 (2001X), EN ISO 374-1 Type B (JKL), EN ISO 374-5, EN 421
- Food contact approved:
- +
- +
- +
Frequently Bought Together