Order Entry
Denmark
ContactUsLinkComponent
Gas management system
Gas management system
Catalog # REST21999
Supplier:  Restek
undefined
Gas management system
Catalog # REST21999
Supplier:  Restek

Specifications

  • For:
    GC
  • Description:
    Gas management system with fittings for 1/8" and 1/4" gas line
  • Seq:
    01
  • W×D×H:
    305×76×356 mm
  • Filter:
    Hydrocarbon
  • Pk:
    1

Specifications

About this item

This gas management system removes moisture, hydrocarbons, and oxygen from carrier gas, extending column lifetime.

  • Produces high-purity carrier gas for most applications
  • Replacing traps is safe and easy
  • Maximum flow: 150 ml/min for optimal performance
Delivery: System includes one each moisture, hydrocarbon and indicating oxygen trap.