Order Entry
Denmark
ContactUsLinkComponent
 
Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution, AVS TITRINORM volumetric solution
  5045.9010
 :  
undefined
Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution, AVS TITRINORM volumetric solution
  5045.9010
 :  

 

  • Pack type:
    Plastic drum
  • Pk:
    10 L
  • Seq:
    5214

 

 

  • Titer:
    0,2185 - 0,2415 mol/l

 

 

 :
 :56.11 g/mol
 :1.009 g/cm³ (20 °C)
 :Ambient
 :0
 :1814
 :8,III
 :GHS05
 :1310-58-3
 :KOH
 :MFCD00003553