Anti-CD163L1 Rabbit Polyclonal Antibody
ABGEAP13979B
: Abgent
- Antistof type:Primary
- Antigen symbol:CD163L1
- Clonality:Polyclonal
- ELISA:Yes
- Vært:Rabbit
- Reactivity:Human
- Western blot:Yes
- Størrelse:100 µg
- Pakn:400 µl
Type: Primary
Antigen: CD163L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human