Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-Histone H2B Sheep Polyclonal Antibody
Anti-Histone H2B Sheep Polyclonal Antibody
Catalog # NOVUNB100-746
Supplier:  Novus Biologicals
Anti-Histone H2B Sheep Polyclonal Antibody
Catalog # NOVUNB100-746
Supplier:  Novus Biologicals

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Histone H2B
  • Clonality:
    Polyclonal
  • Vært:
    Sheep
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Bovine
  • Western blot:
    Yes
  • Størrelse:
    125 μg
  • Pakn:
    125 µG

Specifications

About this item

Sheep Polyclonal Histone H2B Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Bovine.

Type: Primary
Antigen: Histone H2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Bovine