Order Entry
Denmark
ContactUsLinkComponent
Anti-APLP2 Rabbit Polyclonal Antibody
Anti-APLP2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-32814
Supplier:  Novus Biologicals
Anti-APLP2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-32814
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid Precursor-Like Protein 2
  • Antigen symbol:
    APLP2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Antigen synonyms:
    APPH|APP2|Amyloid protein homolog |Amyloid-like protein 2 precursor|CDEI-box binding protein
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit Polyclonal APLP2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: APLP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human