Order Entry
Denmark
ContactUsLinkComponent
 
Anti-MMP2 Mouse Monoclonal Antibody [clone: MMP2/8B4] (Biotin)
  NOVUNB200-114B
Anti-MMP2 Mouse Monoclonal Antibody [clone: MMP2/8B4] (Biotin)
  NOVUNB200-114B

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 2
  • Antigen symbol:
    MMP2
  • Clonality:
    Monoclonal
  • Clone:
    MMP2/8B4
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    4313
  • Antigen synonyms:
    MMP-2|72 kDa gelatinase|matrix metalloproteinase-2|collagenase type IV-A|neutrophil gelatinase|gelatinase A|matrix metalloproteinase-II|72 kDa type IV collagenase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Mouse Monoclonal MMP2 Antibody (MMP2/8B4) [Biotin]. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 2 ?g/ml

Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human