Anti-GOLGA6 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13482R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Golgin Subfamily A Member 6A
- Antigen symbol:GOLGA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:GOG6A_HUMAN|GOLGA6|GOLGA|golgin like protein|golgi autoantigen golgin subfamily a member 6|Golgin linked to PML|Golgi autoantigen golgin subfamily a 6|GOLGA6A|HGNC:13567.|Golgin-like protein|GLP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GOLGA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human