Order Entry
Denmark
ContactUsLinkComponent
Anti-alpha GalCer:CD1d Complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
Anti-alpha GalCer:CD1d Complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
Supplier:  E-Bioscience
Certificates
Anti-alpha GalCer:CD1d Complex Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L363]
Supplier:  E-Bioscience

Specifications

  • Catalog No:
  • BOSSBS-3270R
  • EBIO12-2019-82
  • Antigen symbol:
  • MAP2K1 + MAP2K2
  • alpha GalCer:CD1d Complex
  • Antigen name:
  • Mitogen-activated protein kinase Kinase 1 And 2
  • alpha GalCer:CD1d Complex
  • Antigen synonyms:
  • PRKMK2|MAPKK 2|MAPKK 1|MAP kinase kinase 2|MAPK/ERK kinase 2|Map2k2|dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MEK 1|ERK activator kinase 2|ERK activator kinase 1|MEK2|MKK2|Dual specificity mitogen-activated protein kinase kinase 2|MAP kinase kinase 1|MAPK/ERK kinase 1|mitogen-activated kinase 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • PE (Phycoerythrin)
  • Clone:
  • L363
  • Reactivity:
  • Human, Rat, Mouse
  • Mouse
  • Host:
  • Rabbit
  • Mouse
  • Gene ID:
  • 5604,5605
  • 12479
  • Isotype:
  • IgG
  • IgG2a kappa
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Flow cytometry:
  • Yes
  • Modification:
  • phospho-Ser218/Ser222
  • Immunogen:
  • 180-230/393
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Purified by Protein A
  • Affinity chromatography
  • Concentration:
  • 1 ug/ul
  • 0.2 mg/mL
  • Cross adsorption:
  • No
  • Storage buffer:
  • Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • PBS, 0.1% gelatin, 0.09% sodium azide, pH 7.2
  • Storage temperature:
  • Store at –20 °C for 12 months
  • 4° C, store in dark, DO NOT FREEZE!
  • Shipping temperature:
  • 4 °C

Specifications

Related Information

Frequently Bought Together