About this item
This MAb reacts with a protein of 20-30 kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease.
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 13C4
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Specifications
- Catalog No:
- BNC470147-500
- BNC470147-100
- Antigen symbol:
- UCHL1
- UCHL1
- Antigen name:
- Ubiquitin Carboxyl-Terminal Esterase L1
- Ubiquitin Carboxyl-Terminal Esterase L1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone 13C4
- Clone 13C4
- Reactivity:
- Zebrafish, Pig, Rabbit, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Zebrafish, Pig, Rabbit, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 518731
- 518731
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide