Order Entry
Denmark
ContactUsLinkComponent
Capillary columns, MXT®-20 (Siltek®-treated stainless steel)
Capillary columns, MXT®-20 (Siltek®-treated stainless steel)
Catalog # REST70320
Supplier:  Restek
undefined
Capillary columns, MXT®-20 (Siltek®-treated stainless steel)
Catalog # REST70320
Supplier:  Restek

Specifications

  • Column Format:
    Capillary
  • Film thickness:
    0,25 µm
  • Ø int.:
    0,25 mm
  • Length:
    15 m
  • Phase:
    MXT®-20
  • Pk:
    1

Specifications

About this item

MXT®-20 columns are general purpose columns for volatile compounds, flavor compounds and alcoholic beverages.

  • Low to midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
  • Temperature range up to 340 °C
  • Equivalent to USP G28, G32 phases

The MXT®-20 polymer is synthesized to exacting standards. All residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight mono-modal distribution and extremely low bleed.