Order Entry
Denmark
ContactUsLinkComponent
Genomic RNA purification kits, Fast RNA™ Pro Blue kit, MP Biomedicals
Genomic RNA purification kits, Fast RNA™ Pro Blue kit, MP Biomedicals
Catalog # ICNA116025050
Supplier:  MP Biomedicals
CAS Number:  
undefined
Genomic RNA purification kits, Fast RNA™ Pro Blue kit, MP Biomedicals
Catalog # ICNA116025050
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial RNA
  • Sample type:
    Bacteria
  • Target molecule:
    RNA
  • Description:
    Fast RNA™ Pro Blue Kit, 50×2 ml preps
  • Description:
    Fast RNA™ Pro Blue Kit

Specifications

About this item

FastRNA® Pro Blue Kit is used to isolate total RNA from gram positive and gram negative bacteria. Isolated RNA is suitable for RT-PCR analysis and other downstream applications.

Using the FastPrep®-24 or FastPrep® FP120 instrument, up to 10¹⁰ cells are homogenised by Lysing Matrix B in impact-resistant 2 ml tubes. Total RNA is released into the proprietary, protective RNApro™ Solution.

For research purposes only.

Delivery: Kit contains RNApro™ Solution (1×55 ml), DEPC-treated H₂O (1×15 ml) and Lysing Matrix B (50×2 ml tubes).