Order Entry
Denmark
ContactUsLinkComponent
Diatomaceous earth sorbents
Catalog # REST26033
Supplier:  Restek
CAS Number:  
undefined
Diatomaceous earth sorbents
Catalog # REST26033
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Diatomaceous earth, bulk adsorbent, in 1 kg packs
  • Environmentally Preferable:
Specifications
About this item
Diatomaceous earth is used as a filter aid to improve extraction efficiency of densely packed soils, such as clays. By mixing the sample with diatomaceous earth, recoveries can be improved and excess moisture can be absorbed.
  • Improves extraction efficiency
  • Adsorbs moisture from samples