Anti-NPY5R Goat Polyclonal Antibody (Biotinylated)
EVBIEB06769-B
- Antibody type:Primary
- Antigen name:Neuropeptide Y receptor Y5
- Antigen symbol:NPY5R
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Affinity Purified
- Form:Liquid
- Gene ID:4889
- Antigen synonyms:NPYR5|NPY-Y5 receptor|neuropeptide Y receptor Y5|NPY5-R|Y5 receptor|NPY5R|NPYY5-R
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:50.7
- Sequence:C-GLSNKENRLEEN.
- Storage temperature:+8 °C
- Concentration:0,5 µg/µl
- Immunogen:Neuropeptide Y receptor Y5, Biotinylated
- Size:200 μL
- Pack type:Plastic Vial
- Pk:0,1 mg
Type: Primary
Antigen: NPY5R
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog