About this item
AlbumiNZ™ Low IgG BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality.
- In tan powder format
- To be stored at 4 °C
- Applied in mammalian cell culture, binding and transport studies
This albumin has very low endotoxin and low IgG to simplify downstream purification, is tested for the absence of viruses according to 9CFR and has the highest possible biosafety level. The albumin supports growth of stem cells and increases protein production from hybridoma cells.
It is readily soluble in water and can only be precipitated by high concentrations of neutral salts such as ammonium sulphate. The solution stability of albumin is very good (especially if the solutions are stored aliquoted and frozen). It is readily coagulated by heat. When heated to 50 °C or above, albumin quite rapidly forms hydrophobic aggregates which do not revert to monomers upon cooling.
Specifications
- CAS No.:9048-46-8
- Source:Serum
- Species:Bovine
- Storage conditions:+4 °C
- Form:Powder
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Purity:>97%
- pH:6,5 - 7,5