HPLC silica columns, Viva Silica (USP L3)
Supplier: Restek
Certificates
About this item
These Viva columns are based on a wide pore material designed for optimal large-molecule separations. Base-deactivated spherical silica is useful for normal-phase or HILIC separations. These columns have a narrow distribution around the mean pore size, permitting a larger portion of the silica surface to play a role in the separation.
- Rugged, spherical particles with 300 Å pore size
- High proportion of pore/surface area available to large molecules
- Excellent for separating peptides or proteins
Specifications
- Catalog No:
- REST951050210
- REST951053E
- REST9510515
- REST9510571
- REST9510572
- REST9510572-700
- REST9510532
- REST951057E
- REST9510532-700
- REST9510533-700
- REST9510535
- REST9510535-700
- REST9510525
- REST9510565
- REST9510525-700
- REST9510565-700
- REST951052E
- REST951056E
- REST9510531
- REST9510575-700
- REST951050212
- REST9510552-700
- REST9510512
- REST9510553-700
- REST9510512-700
- REST9510555
- REST951051E
- REST9510555-700
- REST9510521
- REST951055E
- REST9510522
- REST9510563-700
- REST9510561
- REST9510522-700
- REST9510575
- REST9510573-700
- REST9510562
- REST9510523-700
- REST9510562-700
- REST9510513-700
- REST9510515-700
- REST9510551
- REST9510552
- REST9510511
- Endcapped:
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3