Order Entry
Denmark
ContactUsLinkComponent
pH/mV/ISE/°C meters, bench top, Orion Star™ A214
pH/mV/ISE/°C meters, bench top, Orion Star™ A214
Supplier:  Thermo Orion
Certificates
undefined
pH/mV/ISE/°C meters, bench top, Orion Star™ A214
Supplier:  Thermo Orion
Specifications
  • Catalog No:
  • 665-0269
  • 665-0270
  • 665-0479
  • 665-0477
  • 665-0478
  • Model:
  • A214
  • A214
  • A214
  • A214
  • A214
  • pH range:
  • –2,000...+20,000
  • –2,000...+20,000
  • –2,000...+20,000
  • –2,000...+20,000
  • –2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or 0,05% of reading (whichever is greater)
  • ±0,2 mV or 0,05% of reading (whichever is greater)
  • ±0,2 mV or 0,05% of reading (whichever is greater)
  • ±0,2 mV or 0,05% of reading (whichever is greater)
  • ±0,2 mV or 0,05% of reading (whichever is greater)
  • ISE range:
  • 0,0001 - 19900
  • 0,0001 - 19900
  • 0,0001 - 19900
  • 0,0001 - 19900
  • 0,0001 - 19900
  • ISE resolution:
  • 0,0001 minimum, auto ranging, up to 3 significant digits (user selectable)
  • 0,0001 minimum, auto ranging, up to 3 significant digits (user selectable)
  • 0,0001 minimum, auto ranging, up to 3 significant digits (user selectable)
  • 0,0001 minimum, auto ranging, up to 3 significant digits (user selectable)
  • 0,0001 minimum, auto ranging, up to 3 significant digits (user selectable)
  • ISE accuracy:
  • ±0,2 or ±0,05% of reading (whichever is greater)
  • ±0,2 or ±0,05% of reading (whichever is greater)
  • ±0,2 or ±0,05% of reading (whichever is greater)
  • ±0,2 or ±0,05% of reading (whichever is greater)
  • ±0,2 or ±0,05% of reading (whichever is greater)
  • ISE calibration:
  • Up to 5 points
  • Up to 5 points
  • Up to 5 points
  • Up to 5 points
  • Up to 5 points
  • ISE units:
  • ppm, M, mg/l, %, ppb, none
  • ppm, M, mg/l, %, ppb, none
  • ppm, M, mg/l, %, ppb, none
  • ppm, M, mg/l, %, ppb, none
  • ppm, M, mg/l, %, ppb, none
  • Temperature range:
  • –5,0...+105,0 °C (22,0...221,0 °F)
  • –5,0...+105,0 °C (22,0...221,0 °F)
  • –5,0...+105,0 °C (22,0...221,0 °F)
  • –5,0...+105,0 °C (22,0...221,0 °F)
  • –5,0...+105,0 °C (22,0...221,0 °F)
  • Temperature resolution:
  • 0,1 °C, 0,1 °F
  • 0,1 °C, 0,1 °F
  • 0,1 °C, 0,1 °F
  • 0,1 °C, 0,1 °F
  • 0,1 °C, 0,1 °F
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Temperature calibration:
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • Temperature compensation:
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Manual or automatic with ATC temperature probe
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery type:
  • 4 AA (sold separately)
  • 4 AA (sold separately)
  • 4 AA (sold separately)
  • 4 AA (sold separately)
  • 4 AA (sold separately)
  • Battery life:
  • 800 hours
  • 800 hours
  • 800 hours
  • 800 hours
  • 800 hours
  • Power supply:
  • Universal AC adapter (included) or 4 AA batteries (sold separately)
  • Universal AC adapter (included) or 4 AA batteries (sold separately)
  • Universal AC adapter (included) or 4 AA batteries (sold separately)
  • Universal AC adapter (included) or 4 AA batteries (sold separately)
  • Universal AC adapter (included) or 4 AA batteries (sold separately)
  • Compliance:
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • Operating temperature range:
  • 5...45 °C (41...113 °F) (ambient)
  • 5...45 °C (41...113 °F) (ambient)
  • 5...45 °C (41...113 °F) (ambient)
  • 5...45 °C (41...113 °F) (ambient)
  • 5...45 °C (41...113 °F) (ambient)
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 2000 with date and time stamp
  • 2000 with date and time stamp
  • 2000 with date and time stamp
  • 2000 with date and time stamp
  • 2000 with date and time stamp
  • Memory:
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • IP rating:
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • IP 54
  • Connections:
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Outputs:
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • Input:
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
  • Alarm:
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • Calibration:
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • Calibration points:
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • W×D×H:
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • 180×240×110 mm
  • Weight:
  • 0,9 kg (2 lbs.)
  • 0,9 kg (2 lbs.)
  • 0,9 kg (2 lbs.)
  • 0,9 kg (2 lbs.)
  • 0,9 kg (2 lbs.)