D-(+)-Glucose, anhydrous, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH (10% aq soln) :4,6 - 6,5
- Melting Point:150 - 170° C
- Loss on Drying:≤0,5% max
- Optical Rotation (c=10, H₂O + 1 drop NH₄OH):Report result
Specification Test Results
Frequently Bought Together