D-(+)-Maltose monohydrate ~99%, white powder
:6363-53-7
:
:C₁₂H₂₂O₁₁·1H₂O
:MFCD00149343
:360.32 g/mol
- Identity Test:Passes
- Purity :~99%
- FTIR:Conforms to Standard
- Moisture:≤6%
- Optical Rotation (c=4, H2O, NH3OH):130° ± 5°
Frequently Bought Together