Ordreindtastning
Denmark
ContactUsLinkComponent
 
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG), crystalline
 
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG), crystalline

 

  • Identity test:
    Passes
  • FTIR:
    Conforms to standard
  • Optical Rotation (c=1, H₂O):
    –66° ± 7°
  • Melting Point:
    180 - 195 °C
  • UV/VIS: λmax (H₂O):
    262 ±3 nm

 

Frequently Bought Together