Thiamine hydrochloride, powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
CAS Number:67-03-8
:
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780
Molecular Weight:337.27 g/mol
Melting Pt:252
Merck Index:13,09366
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH (1% aq soln):2,7 - 3,4
- OD400 (1% aq soln):≤0.25
- Loss on Drying:≤5,0%
- Residue on Ignition:≤0,2%
Specification Test Results
Frequently Bought Together