Ganglioside GQ1b, Tetrasialo, Tetraammonium Salt (from bovine brain) ≥98% (by TLC), Millipore®
Catalog # 345754-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 µG
- Seq:1286
Specifications
Specification Test Results
- Synonyms:IV3(Neu5Ac)2II3(Neu5Ac)2-GgOse4, 4NH4
- Purity:≥98% by TLC
- Ship Code:Ambient Temperature only
- Toxicity:Irritant
- Storage:−20 °C
- Do not freeze:Ok to freeze
- Form:Lyophilized
- Solubility:Chloroform:Methanol (2:1) plus a few drops of H₂O. Will form a micellar solution with H2O
- Special Instructions:Following reconstitution, aliquot and freeze (−20 °C). Stock solutions are stable for up to 3 months at −20 °C
Specification Test Results
About this item
:
Formula:C₁₀₆H₁₇₈N₆O₅₅.4NH₄
MDL Number:MFCD02098588