Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose, powder

D-(-)-Fructose, powder

500 g
Catalog # ICNA02102186.5
Supplier:   MP Biomedicals
D-(-)-Fructose, powder
Catalog # ICNA02102186.5
Supplier:  MP Biomedicals
CAS Number:  57-48-7
D-(-)-Fructose, powder
D-(-)-Fructose, powder

500 g

CAS Number:  57-48-7
Price
365,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Fructose or fruit sugar, is a simple monosaccharide found in many plants. It is one of the three dietary monosaccharides, along with glucose and galactose, that are absorbed directly into the bloodstream during digestion. Fructose is a 6-carbon polyhydroxyketone. It is an isomer of glucose which can be anaerobically fermented by yeast or bacteria.

  • Presentation: Powder

Fructose and other monosaccharides are used for the diagnosis of cancer as cancer cells have shown a higher level of fructose utilisation as compared to glucose.

Soluble in water and alcohols.

Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Melting Pt:110…120 °C
  • Density:1.6 g/mL (lit.)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Optical Rotation (c=2, water):
    -88° to -98°
  • Arsenic:
    <1 ppm
  • Heavy Metals:
    <5 ppm
  • Moisture (KF):
    0.02%

Specifications

  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.