Order Entry
Denmark
ContactUsLinkComponent
Test tube holder, stainless steel/wooden handle
Test tube holder, stainless steel/wooden handle
Catalog # BOCH2283
Leverandør:  BOCHEM
undefined
Test tube holder, stainless steel/wooden handle
Catalog # BOCH2283
Leverandør:  BOCHEM
Specifikationer
  • Length:
    250 mm
  • Pk:
    1
  • Spanning area:
    10 - 25 mm
Specifikationer
Om denne vare
This test tube holder is made from stainless steel (18/10) and features a wooden handle. For tubes with a diameter from 10 to 25 mm.