Anti-C16orf9 Mouse Polyclonal Antibody
Catalog # ABNOH00083986-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:C16orf9
- Clonality:Polyclonal
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:Full-length
- Gene ID:83986
- Sequence:MLDHKDLEAEIHPLKNEERKSQENLGNPSKNEDNVKSAPPQSRLSRCRAAAFFLSLFLCLFVVFVVSFVIPCPDRPASQRMWRIDYSAAVIYDFLAVDDINGDRIQDVLFLYKNTNSSNNFSRSCVDEGFSSPCTFAAAVSGANGSTLWERPVAQDVALVECAVPQPRGSEAPSACILVGRPSSFIAVNLFTGETLWNHSSSFSGNASILSPLLQVPDVDGDGAPDLLVLTQEREEVSGHLYSGSTGHQIGLRGSLGVDGESGFLLHVTRTGAHYILFPCASSLCGCSVKGLYEKVTGSGGPFKSDPHWESMLNATTRRMLSHSSGAVRYLMHVPGNAGADVLLVGSEAFVLLDGQELTPRWTPKAAHVLRKPIFGRYKPDTLAVAVENGTGTDRQILFLDLGTGAVLCSLALPSLPGGPLSASLPTADHRSAFFFWGLHELGSTSETETGEARHSLYMFHPTLPRVLLELANVSTHIVAFDAVLFEPSRHAAYILLTGPADSEAPGLVSVIKHKVRDLVPSSRVVRLGEGGPDSDQAIRDRFSRLRYQSEA
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human C16orf9 protein.
Type: Primary
Antigen: C16orf9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype:
Reactivity: Human,Rat