Anti-PTPRCC5 Mouse Monoclonal Antibody [clone: 135-4C5]
ABNOMAB13413
: Abnova
- Antibody type:Primary
- Antigen symbol:PTPRCC5
- Clonality:Monoclonal
- Clone:135-4C5
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Gene ID:5788
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against native human PTPRC.
Type: Primary
Antigen: PTPRCC5
Clonality: Monoclonal
Clone: 135-4C5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human