Anti-S100A9/840 Mouse Monoclonal Antibody [clone: MRP14/840]
ABNOMAB13429
: Abnova
- Antibody type:Primary
- Antigen symbol:S100A9/840
- Clonality:Monoclonal
- Clone:MRP14/840
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:6280
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human S100A9.
Type: Primary
Antigen: S100A9/840
Clonality: Monoclonal
Clone: MRP14/840
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human