Anti-BCL2796 Mouse Monoclonal Antibody [clone: BCL2/796]
ABNOMAB13419
: Abnova
- Antibody type:Primary
- Antigen symbol:BCL2796
- Clonality:Monoclonal
- Clone:BCL2/796
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:596
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human BCL2.
Type: Primary
Antigen: BCL2796
Clonality: Monoclonal
Clone: BCL2/796
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human