Anti-PECAM10 Mouse Monoclonal Antibody [clone: C31.10]
ABNOMAB13395
: Abnova
- Antibody type:Primary
- Antigen symbol:PECAM10
- Clonality:Monoclonal
- Clone:C31.10
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:5175
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human PECAM1.
Type: Primary
Antigen: PECAM10
Clonality: Monoclonal
Clone: C31.10
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human